missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxytamoxifen, MedChemExpress

Product Code. 16467218
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16467218

Brand: Medchem Express HY16950

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 4-Hydroxytamoxifen ((Z)-4-Hydroxytamoxifen) is an orally active, selective estrogen receptor modulator (SERM). 4-Hydroxytamoxifen ((Z)-4-Hydroxytamoxifen) induces CRISPR/Cas9 systems based on ER mediated nucleus translocation.

4-Hydroxytamoxifen ((Z)-4-Hydroxytamoxifen) is an orally active, selective estrogen receptor modulator (SERM). 4-Hydroxytamoxifen ((Z)-4-Hydroxytamoxifen) induces CRISPR/Cas9 systems based on ER mediated nucleus translocation.

Specifications

Chemical Name or Material 4-Hydroxytamoxifen
CAS 68047-06-3
Color White
Purity Grade Notes Research
Molecular Formula C26H29NO2
Quantity 10 mg
Synonym (Z)-4-Hydroxytamoxifen trans-4-Hydroxytamoxifen (Z)-Afimoxifene
Solubility Information DMSO : 50 mg/mL (129.03 mM; ultrasonic and warming and heat to 60°C) ∣Ethanol : 20 mg/mL (51.61 mM; Need ultrasonic)
SMILES CC/C(C1=CC=CC=C1)=C(C2=CC=C(OCCN(C)C)C=C2)\C3=CC=C(O)C=C3.[(Z)]
Molecular Weight (g/mol) 387.51
Formula Weight 387.51
Percent Purity 99.5%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.