missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxythiophenol, 97%

Product Code. 11381577
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381577 25 g 25g
11371577 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381577 Supplier Thermo Scientific Alfa Aesar Supplier No. L04429.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 637-89-8 | C6H6OS | 126.17 g/mol

4-Hydroxythiophenol is used to improve charge separation in PbSe quantum dots (QDs) as sensitizers for TiO2. It helps in the activation of neocarzinostatin-chromophore. Also used as a resin monomer in the study of the different gold nanocrystal core-resin shell structures

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxythiophenol is used to improve charge separation in PbSe quantum dots (QDs) as sensitizers for TiO2. It helps in the activation of neocarzinostatin-chromophore. Also used as a resin monomer in the study of the different gold nanocrystal core-resin shell structures

Solubility
Soluble in water.

Notes
Air Sensitive. Store at 4°C. Incompatible with air and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-89-8
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.17
MDL Number MFCD00004850
InChI Key BXAVKNRWVKUTLY-UHFFFAOYSA-N
Synonym 4-mercaptophenol, 4-hydroxythiophenol, 4-hydroxybenzenethiol, p-hydroxythiophenol, phenol, 4-mercapto, p-mercaptophenol, thiohydroquinone, monothiohydroquinone, 4-hydroxy thiophenol, hydroquinone, monothio
PubChem CID 240147
IUPAC Name 4-sulfanylphenol
SMILES OC1=CC=C(S)C=C1

Specifications

Melting Point 33°C to 37°C
Density 1.255
Boiling Point 256°C
Flash Point 110°C (230°F)
Odor Unpleasant
Quantity 25 g
UN Number UN1759
Beilstein 2039306
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 126.18
Percent Purity 97%
Chemical Name or Material 4-Hydroxythiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.