missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-1-trityl-1H-imidazole, 98%, Thermo Scientific Chemicals

Product Code. 15569583 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15569583 5 g 5g
15559583 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15569583 Supplier Thermo Scientific Acros Supplier No. 459200050
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96797-15-8
Molecular Formula C22H17IN2
Molecular Weight (g/mol) 436.30
MDL Number MFCD02179542
InChI Key DXJZJYPLPZEYBH-UHFFFAOYSA-N
Synonym 4-iodo-1-trityl-1h-imidazole, 4-iodo-1-triphenylmethyl imidazole, 1-trityl-4-iodoimidazole, 4-iodo-1-triphenylmethyl-1h-imidazole, 4-iodo-1-trityl-imidazole, 1h-imidazole, 4-iodo-1-triphenylmethyl, pubchem8995, 4-iodo-l-tritylimidazole, n1-trityl-4-iodoimidazole
PubChem CID 618252
IUPAC Name 4-iodo-1-tritylimidazole
SMILES IC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 223.0°C to 227.5°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 436.3
Percent Purity 98%
Chemical Name or Material 4-Iodo-1-trityl-1H-imidazole
compliance-icons
Product Identifier
  • 4-Iodo-1-trityl-1H-imidazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.