missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-2,6-dimethylphenol, 97%

Product Code. 11458687
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458687 2.5 g 2.5g
11468687 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11458687 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. A19840.88
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

CAS: 10570-67-9 | C8H9IO | 248.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 10570-67-9
Molecular Formula C8H9IO
Molecular Weight (g/mol) 248.06
MDL Number MFCD00094415
InChI Key HUUNIMCCAGNBDF-UHFFFAOYSA-N
Synonym 2,6-dimethyl-4-iodophenol, 4-iodo-2,6-dimethyl-phenol, 4-iodo-2,6-xylenol, acmc-2098hg, phenol,4-iodo-2,6-dimethyl, huunimccagnbdf-uhfffaoysa, phenol, 4-iodo-2,6-dimethyl
PubChem CID 555318
IUPAC Name 4-iodo-2,6-dimethylphenol
SMILES CC1=CC(I)=CC(C)=C1O

Tekniske data

Melting Point 96°C to 101°C
Quantity 2.5 g
Sensitivity Light sensitive
Formula Weight 248.06
Percent Purity 97%
Chemical Name or Material 4-Iodo-2,6-dimethylphenol

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.