missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-2-nitrotoluene, 97%

Product Code. 11458527
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458527 5 g 5g
11468527 25 g 25g
11478527 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458527 Supplier Thermo Scientific Alfa Aesar Supplier No. A18607.06
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 41252-97-5 | C7H6INO2 | 263.034 g/mol

4-Iodo-2-nitrotoluene is used to produce 3-(4-methyl-3-nitro-phenyl)-1H-indole at the temperature of 40°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodo-2-nitrotoluene is used to produce 3-(4-methyl-3-nitro-phenyl)-1H-indole at the temperature of 40°C.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41252-97-5
Molecular Formula C7H6INO2
Molecular Weight (g/mol) 263.034
MDL Number MFCD00051090
InChI Key QLMRDNPXYNJQMQ-UHFFFAOYSA-N
Synonym 4-iodo-2-nitrotoluene, benzene, 4-iodo-1-methyl-2-nitro, pubchem15938, 2-nitro-4-iodotoluene, acmc-1apfk, maybridge1_008109, ksc493s9d, 4-iodo-1-methyl-2-nitro-benzene
PubChem CID 170481
IUPAC Name 4-iodo-1-methyl-2-nitrobenzene
SMILES CC1=C(C=C(C=C1)I)[N+](=O)[O-]

Specifications

Melting Point 58°C to 62°C
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 263.03
Percent Purity 97%
Chemical Name or Material 4-Iodo-2-nitrotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.