missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-3-methylaniline, 98%

Product Code. 10603573 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10603573 5 g Glass bottle 5g
10282752 25 g Glass bottle 25g
10433595 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10603573 Supplier Thermo Scientific Acros Supplier No. 357800050
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4949-69-3 | C7H8IN | 233.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4949-69-3
MDL Number MFCD01569451
InChI Key UISBOJCPTKUBIC-UHFFFAOYSA-N
Synonym 3-methyl-4-iodoaniline, 2-iodo 5-aminotoluene, 4-iodo-3-methyl aniline, acmc-1ak0z, benzenamine,4-iodo-3-methyl
PubChem CID 2734275
IUPAC Name 4-iodo-3-methylaniline
SMILES CC1=C(C=CC(=C1)N)I

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 50°C to 55°C
Color Beige-Brown to Gray
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H8IN
Quantity 5 g
Molecular Weight (g/mol) 233.05
Formula Weight 233.05
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodo-3-methylaniline
Show More Show Less
compliance-icons
Product Identifier
  • 4-Iodo-3-methylaniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.