missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-3-nitroanisole, 98+%

Product Code. 11332587
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332587 25 g 25g
11322587 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332587 Supplier Thermo Scientific Alfa Aesar Supplier No. L11714.14
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 58755-70-7 | C7H6INO3 | 279.033 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58755-70-7
Molecular Formula C7H6INO3
Molecular Weight (g/mol) 279.033
MDL Number MFCD00024195
InChI Key JWZODIRSJJQOKY-UHFFFAOYSA-N
Synonym 4-iodo-3-nitroanisole, 3-nitro-4-iodoanisole, 2-iodo-5-methoxynitrobenzene, 1-iodo-4-methoxy-2-nitro-benzene, benzene, 1-iodo-4-methoxy-2-nitro, 3-nitro-4-iodoanisol, acmc-1b13g, ksc494s9p, 1-iodo-2-nitro-4-methoxybenzene, attercop-chm at115569
PubChem CID 292659
IUPAC Name 1-iodo-4-methoxy-2-nitrobenzene
SMILES COC1=CC(=C(C=C1)I)[N+](=O)[O-]

Specifications

Melting Point 60°C to 63°C
Quantity 25 g
Beilstein 2365592
Sensitivity Light sensitive
Formula Weight 279.04
Percent Purity ≥98%
Chemical Name or Material 4-Iodo-3-nitroanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.