missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-3-nitrotoluene, 97%, Thermo Scientific™

Product Code. 10459052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
Product Code. Quantity Packaging unitSize
10459052 5 g Glass bottle 5g
1 options
This item is not returnable. View return policy

Product Code. 10459052

Brand: Thermo Scientific Acros 326300050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5326-39-6
CAS Min % 96.0
CAS Max % 100.0
Melting Point 52°C to 54°C
Color Orange to Brown
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C7H6INO2
MDL Number MFCD00024196
Quantity 5 g
Synonym 4-iodo-3-nitrotoluene, benzene, 1-iodo-4-methyl-2-nitro, 3-nitro-4-iodotoluol, 3-nitro-4-iodotoluene, pubchem4550, acmc-209l5f, ksc597g8t, 4-jod-3-nitro-1-methylbenzol, 1-iodo-4-methyl-2-nitro-benzene
InChI Key DKYCDXAZCHMPSJ-UHFFFAOYSA-N
SMILES CC1=CC=C(I)C(=C1)[N+]([O-])=O
IUPAC Name 1-iodo-4-methyl-2-nitrobenzene
Molecular Weight (g/mol) 263.03
PubChem CID 138455
Formula Weight 263.03
Percent Purity 97%
Physical Form Glistening Powder
Chemical Name or Material 4-Iodo-3-nitrotoluene, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.