missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzoic acid, 97%

Product Code. 11469602
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469602 250 g 250g
11449602 10 g 10g
11459602 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469602 Supplier Thermo Scientific Alfa Aesar Supplier No. A10730.30
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 619-58-9 | C7H5IO2 | 248.02 g/mol

4-Iodobenzoic acid was used in the synthesis of [hydroxy(4-carboxyphenyl)iodonium]ion in situ that helps in the cleavage of a variety of alkenes. It is is used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzoic acid was used in the synthesis of [hydroxy(4-carboxyphenyl)iodonium]ion in situ that helps in the cleavage of a variety of alkenes. It is is used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

Solubility
Partly miscible with water. Soluble in alcohol & ether.

Notes
It is sensitive to light. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-58-9
Molecular Formula C7H5IO2
Molecular Weight (g/mol) 248.02
MDL Number MFCD00002533
InChI Key GHICCUXQJBDNRN-UHFFFAOYSA-N
Synonym p-iodobenzoic acid, benzoic acid, 4-iodo, benzoic acid, p-iodo, 4-iodo-benzoic acid, p-iodobenzenecarboxylic acid, 4-jodbenzoesaeure, unii-ipo4lyq1en, 4-jodbenzoesaeure german, 4-iodo benzoic acid, ipo4lyq1en
PubChem CID 12085
IUPAC Name 4-iodobenzoic acid
SMILES OC(=O)C1=CC=C(I)C=C1

Specifications

Melting Point ∼265°C (decomposition)
Density 2.18
Quantity 250 g
Beilstein 1860232
Sensitivity Light sensitive
Solubility Information Partly miscible with water. Soluble in alcohol & ether.
Formula Weight 248.02
Percent Purity 97%
Chemical Name or Material 4-Iodobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.