missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzoyl chloride, 98%

Product Code. p-7028206
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g

Product Code. 11467066

Brand: Thermo Scientific Alfa Aesar B25388.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

CAS: 1711-02-0 | C7H4ClIO | 266.462 g/mol

4-Iodobenzoyl chloride is employed in the preparation of pyrroles by reaction with imines and acetylenes mediated by isocyanides using palladium catalysis. It is also used in the preparation of N-(1-benzylpyrrolidin-3-yl)arylbenzamides, potential human dopamine D4 antagonists. Further, it is used to prepare [2]rotaxane monomer and poly[2] rotaxane. In addition to this, it is important in the modification of poly(allylamine) in order to make the polymer X-ray visible.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzoyl chloride is employed in the preparation of pyrroles by reaction with imines and acetylenes mediated by isocyanides using palladium catalysis. It is also used in the preparation of N-(1-benzylpyrrolidin-3-yl)arylbenzamides, potential human dopamine D4 antagonists. Further, it is used to prepare [2]rotaxane monomer and poly[2] rotaxane. In addition to this, it is important in the modification of poly(allylamine) in order to make the polymer X-ray visible.

Solubility
Soluble in toluene and benzene.

Notes
Moisture sensitive. Incompatible with strong bases and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1711-02-0
Molecular Formula C7H4ClIO
Molecular Weight (g/mol) 266.462
MDL Number MFCD00001058
InChI Key NJAKCIUOTIPYED-UHFFFAOYSA-N
Synonym p-iodobenzoyl chloride, benzoyl chloride, 4-iodo, benzoyl chloride, p-iodo, 4-iodobenzoic acid chloride, 4-iodobenzoylchloride, 4-iodo-benzoyl chloride, 4iodobenzoyl chloride, 4-iodo-benzoylchlorid, rho-iodobenzoyl chloride, 4-iodo benzoyl chloride
PubChem CID 74373
IUPAC Name 4-iodobenzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)I

Specifications

Melting Point 61°C to 65°C
Boiling Point 120°C to 121°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 774718
Sensitivity Light and moisture sensitive
Solubility Information Soluble in toluene and benzene.
Formula Weight 266.47
Percent Purity 98%
Chemical Name or Material 4-Iodobenzoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Iodobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 4-Iodobenzoyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.