missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobiphenyl, 97%

Product Code. 11397309
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397309 25 g 25g
11377309 1 g 1g
11387309 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397309 Supplier Thermo Scientific Alfa Aesar Supplier No. H27453.14
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1591-31-7 | C12H9I | 280.11 g/mol

4-Iodobiphenyl is used as a starting material for the preparation of p-terphenyl derivatives. It is also used in Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobiphenyl is used as a starting material for the preparation of p-terphenyl derivatives. It is also used in Suzuki reaction.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1591-31-7
Molecular Formula C12H9I
Molecular Weight (g/mol) 280.11
MDL Number MFCD00019028
InChI Key NXYICUMSYKIABQ-UHFFFAOYSA-N
Synonym 4-iodobiphenyl, 4-iodo-1,1'-biphenyl, 1,1'-biphenyl, 4-iodo, p-iodobiphenyl, 4-iododiphenyl, p-phenyliodobenzene, 4-biphenyl iodide, biphenyl, 4-iodo, 1-iodo-4-phenyl-benzene, 4-iodo-1-phenylbenzene
PubChem CID 15322
IUPAC Name 1-iodo-4-phenylbenzene
SMILES IC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 110°C to 114°C
Quantity 25 g
UN Number UN3077
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 280.11
Percent Purity 97%
Chemical Name or Material 4-Iodobiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.