missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobutyl acetate, 96%, stab. with copper

Product Code. 15474467
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15474467 10 g 10g
15484467 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474467 Supplier Thermo Scientific Alfa Aesar Supplier No. H31826.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40596-44-9 | C6H11IO2 | 242.056 g/mol

4-Iodobutyl acetate is used in the preparation of tricyclohexyl(acetoxybutyl))phosphonium iodide, tri-n-hexyl(acetoxybutyl))phosphonium iodide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobutyl acetate is used in the preparation of tricyclohexyl(acetoxybutyl))phosphonium iodide, tri-n-hexyl(acetoxybutyl))phosphonium iodide

Solubility
Difficult to mix.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40596-44-9
Molecular Formula C6H11IO2
Molecular Weight (g/mol) 242.056
MDL Number MFCD00010664
InChI Key FLPVVTDEKLGZDZ-UHFFFAOYSA-N
Synonym tech., 4-iodobutylacetate, 4-acetoxybutyl iodide, 4-iodo-butyl acetate, acmc-20amg9, acetic acid 4-iodobutyl ester, acetic acid 4-iodo-butyl ester, 1-butanol, 4-iodo-,1-acetate, 4-iodobutyl acetate, technical grade
PubChem CID 2733405
IUPAC Name 4-iodobutyl acetate
SMILES CC(=O)OCCCCI

Specifications

Density 1.61
Boiling Point 94°C to 95°C (5.5 mmHg)
Flash Point 91°C (196°F)
Linear Formula CH3CO2(CH2)4I
Refractive Index 1.5115
Quantity 10 g
Sensitivity Light sensitive
Solubility Information Difficult to mix.
Formula Weight 242.05
Percent Purity 96%
Chemical Name or Material 4-Iodobutyl acetate, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.