missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenol, TRC

Product Code. 30137564
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137564 50 g 50g
30004943 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137564 Supplier Toronto Research Chemicals Supplier No. TRCI71751050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-38-5
Molecular Formula C6H5IO
Molecular Weight (g/mol) 220.01
Synonym p-Iodophenol, 1-Iodo-4-hydroxybenzene, 4-Hydroxyiodobenzene, 4-Hydroxyphenyl Iodide, 4-Iodophenol, NSC 91464, p-Hydroxyiodobenzene, p-Iodophenol
SMILES Oc1ccc(I)cc1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H5IO/c7-5-1-3-6(8)4-2-5/h1-4,8H
Formula Weight 219.9385
Chemical Name or Material 4-Iodophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.