missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenoxyacetic acid, 98+%, Thermo Scientific™

Product Code. 15498789
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15498789 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15498789 Supplier Thermo Scientific Alfa Aesar Supplier No. L01196.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1878-94-0
Melting Point 156°C to 158°C
Molecular Formula C8H7IO3
MDL Number MFCD00014359
Quantity 5 g
Beilstein 2259999
Sensitivity Light sensitive
Synonym 4-iodophenoxyacetic acid, 2-4-iodophenoxy acetic acid, 4-iodophenoxy acetic acid, p-iodophenoxyacetic acid, p-iodophenoxy acetic acid, acetic acid, 4-iodophenoxy, 4-iodophenoxyacetic, 4-iodophenoxyaceticacide, acmc-209eqm, 4-iodo phenoxyacetic acid
InChI Key RKORKXFKXYYHAQ-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1OCC(=O)O)I
IUPAC Name 2-(4-iodophenoxy)acetic acid
Molecular Weight (g/mol) 278.045
PubChem CID 74658
Formula Weight 278.05
Percent Purity ≥98%
Chemical Name or Material 4-Iodophenoxyacetic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.