missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenyl isocyanate, 97%

Product Code. 11352847
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352847 5 g 5g
11342847 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352847 Supplier Thermo Scientific Alfa Aesar Supplier No. L16000.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 15845-62-2 | C7H4INO | 245.019 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15845-62-2
Molecular Formula C7H4INO
Molecular Weight (g/mol) 245.019
MDL Number MFCD00013875
InChI Key KVMKUTIUJGYHBA-UHFFFAOYSA-N
Synonym 4-iodophenyl isocyanate, 4-iodophenylisocyanate, 1-iodo-4-isocyanato-benzene, p-iodophenylisocyanate, acmc-1bsbj, benzene,1-iodo-4-isocyanato
PubChem CID 3399787
IUPAC Name 1-iodo-4-isocyanatobenzene
SMILES C1=CC(=CC=C1N=C=O)I

Specifications

Melting Point 43°C to 47°C
Boiling Point 84°C (5 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Quantity 5 g
UN Number UN2811
Beilstein 2716151
Sensitivity Light and moisture sensitive
Formula Weight 245.02
Percent Purity 97%
Chemical Name or Material 4-Iodophenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.