missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenylhydrazine, 95%

Product Code. 11393686
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393686 5 g 5g
11383686 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393686 Supplier Thermo Scientific Alfa Aesar Supplier No. L14806.06
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13116-27-3 | C6H7IN2 | 234.04 g/mol

It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Light Sensitive, store in dark. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13116-27-3
Molecular Formula C6H7IN2
Molecular Weight (g/mol) 234.04
MDL Number MFCD00052688
InChI Key IQMLCMKMSBMMGR-UHFFFAOYSA-N
Synonym 4-iodophenyl hydrazine, 1-4-iodophenyl hydrazine, 4-iodo-phenyl-hydrazine, 4-iodo phenyl hydrazine, p-iodophenyl-hydrazine, 4-iodophenyl diazane, pubchem15797, acmc-1brf1, 1-hydrazino-4-iodobenzene, 4-iodophenylhydrazine
PubChem CID 704591
IUPAC Name (4-iodophenyl)hydrazine
SMILES C1=CC(=CC=C1NN)I

Specifications

Melting Point 99°C to 103°C
Quantity 5 g
Beilstein 742475
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 234.04
Percent Purity 95%
Chemical Name or Material 4-Iodophenylhydrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.