missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylanisole, 95%

Product Code. 11438806
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438806 25 g 25g
11448806 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11438806 Supplier Thermo Scientific Alfa Aesar Supplier No. A16970.14
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4132-48-3 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 4132-48-3
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00015033
InChI Key JULZQKLZSNOEEJ-UHFFFAOYSA-N
Synonym 4-isopropylanisole, 1-isopropyl-4-methoxybenzene, 4-methoxycumene, p-isopropylanisole, anisole, p-isopropyl, 2-4-methoxyphenyl propane, benzene, 1-methoxy-4-1-methylethyl, 1-methoxy-4-propan-2-yl benzene, p-methoxycumene, p-methoxyisopropylbenzene
PubChem CID 77783
IUPAC Name 1-methoxy-4-propan-2-ylbenzene
SMILES COC1=CC=C(C=C1)C(C)C

Spezifikation

Density 0.935
Boiling Point 212°C to 213°C
Flash Point 83°C (181°F)
Refractive Index 1.503
Quantity 25 g
Beilstein 2080269
Formula Weight 150.22
Percent Purity 95%
Chemical Name or Material 4-Isopropylanisole

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.