missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylcinnamic acid, predominantly trans, 98+%

Product Code. 11402148
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11402148 5 g 5g
11412148 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11402148 Supplier Thermo Scientific Alfa Aesar Supplier No. A15156.06
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 3368-21-6 | C12H14O2 | 190.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3368-21-6
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD00016544
InChI Key SJDOOXOUSJDYFE-VMPITWQZSA-N
Synonym 4-isopropylcinnamic acid, 3-4-isopropylphenyl acrylic acid, p-isopropylcinnamic acid, 2e-3-4-isopropylphenyl acrylic acid, cinnamic acid, p-isopropyl, 2-propenoic acid, 3-4-1-methylethyl phenyl, 2e-3-4-propan-2-yl phenyl prop-2-enoic acid, e-3-4-isopropylphenyl acrylic acid, e-3-4-isopropyl-phenyl-acrylic acid, 2e-3-4-isopropylphenyl prop-2-enoic acid
PubChem CID 719753
IUPAC Name (E)-3-(4-propan-2-ylphenyl)prop-2-enoic acid
SMILES CC(C)C1=CC=C(\C=C\C(O)=O)C=C1

Specifications

Melting Point 157°C to 161°C
Quantity 5 g
Beilstein 2046679
Formula Weight 190.24
Percent Purity ≥98%
Chemical Name or Material 4-Isopropylcinnamic acid, predominantly trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.