missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylphenol, 98%

Product Code. 11415227
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415227 250 g 250g
11405227 50 g 50g
11324709 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11415227 Supplier Thermo Scientific Alfa Aesar Supplier No. B22620.30
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 99-89-8 | C9H12O | 136.194 g/mol

4-Isopropylphenol was used in the synthesis of vinyl compounds. It is used as a flavor & fragrance agent in cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Isopropylphenol was used in the synthesis of vinyl compounds. It is used as a flavor & fragrance agent in cosmetics.

Solubility
Slightly soluble in water, soluble in ethanol and ethyl ether.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-89-8
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002372
InChI Key YQUQWHNMBPIWGK-UHFFFAOYSA-N
Synonym 4-isopropylphenol, p-isopropylphenol, p-cumenol, australol, phenol, 4-1-methylethyl, 4-1-methylethyl phenol, 1-hydroxy-4-isopropylbenzene, 4-propan-2-yl phenol, phenol, p-isopropyl, para-isopropylphenol
PubChem CID 7465
IUPAC Name 4-propan-2-ylphenol
SMILES CC(C)C1=CC=C(C=C1)O

Specifications

Melting Point 59°C to 61°C
Boiling Point 211°C to 212°C
Flash Point 108°C (226°F)
Quantity 250 g
UN Number UN2430
Beilstein 1363564
Solubility Information Slightly soluble in water,soluble in ethanol and ethyl ether.
Formula Weight 136.2
Percent Purity 98%
Chemical Name or Material 4-Isopropylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.