missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylphenyl isothiocyanate, 96%, Thermo Scientific Chemicals

Product Code. 11327816
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327816 2 g 2g
11337816 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Codice prodotto. 11327816 Fornitore Thermo Scientific Alfa Aesar N. del fornitore L12949.04
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 89007-45-4
Molecular Formula C10H11NS
Molecular Weight (g/mol) 177.27
MDL Number MFCD00022060
InChI Key SNWJJTIJIDXIHL-UHFFFAOYSA-N
Synonym 4-isopropylphenyl isothiocyanate, 1-isopropyl-4-isothiocyanatobenzene, 4-isopropylphenylisothiocyanate, 1-isothiocyanato-4-propan-2-yl benzene, 4-methylethyl benzenisothiocyanate, acmc-20amft, 4isopropylphenylisothiocyanate, 4-iso-propylphenyl isothiocyanate, 1-isopropyl-4-isothiocyanato-benzene, 1-isopropyl-4-isothiocyanatobenzene #
PubChem CID 145194
IUPAC Name 1-isothiocyanato-4-propan-2-ylbenzene
SMILES CC(C)C1=CC=C(C=C1)N=C=S

Specifica

Density 1.03
Boiling Point 154°C to 155°C (20 mmHg)
Refractive Index 1.605
Quantity 2 g
UN Number UN2922
Beilstein 3240731
Sensitivity Moisture sensitive
Formula Weight 177.27
Percent Purity 96%
Chemical Name or Material 4-Isopropylphenyl isothiocyanate

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.