missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylphenyl isothiocyanate, 96%, Thermo Scientific Chemicals

Product Code. 11337816
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337816 10 g 10g
11327816 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337816 Supplier Thermo Scientific Alfa Aesar Supplier No. L12949.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89007-45-4
Molecular Formula C10H11NS
Molecular Weight (g/mol) 177.27
MDL Number MFCD00022060
InChI Key SNWJJTIJIDXIHL-UHFFFAOYSA-N
Synonym 4-isopropylphenyl isothiocyanate, 1-isopropyl-4-isothiocyanatobenzene, 4-isopropylphenylisothiocyanate, 1-isothiocyanato-4-propan-2-yl benzene, 4-methylethyl benzenisothiocyanate, acmc-20amft, 4isopropylphenylisothiocyanate, 4-iso-propylphenyl isothiocyanate, 1-isopropyl-4-isothiocyanato-benzene, 1-isopropyl-4-isothiocyanatobenzene #
PubChem CID 145194
IUPAC Name 1-isothiocyanato-4-propan-2-ylbenzene
SMILES CC(C)C1=CC=C(C=C1)N=C=S

Specifications

Density 1.03
Boiling Point 154°C to 155°C (20 mmHg)
Refractive Index 1.605
Quantity 10 g
UN Number UN2922
Beilstein 3240731
Sensitivity Moisture sensitive
Formula Weight 177.27
Percent Purity 96%
Chemical Name or Material 4-Isopropylphenyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.