missing translation for 'onlineSavingsMsg'
Learn More

4-Mercaptobenzoic acid, 90%

Product Code. 10274974 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10274974 25 g Glass bottle 25g
10264974 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10274974 Supplier Thermo Scientific Acros Supplier No. 373300250
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1074-36-8 | C7H6O2S | 154.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1074-36-8
Molecular Formula C7H6O2S
Molecular Weight (g/mol) 154.18
MDL Number MFCD00016617
InChI Key LMJXSOYPAOSIPZ-UHFFFAOYSA-N
Synonym 4-mercaptobenzoic acid, 4-mercaptobenzoate, benzoic acid, 4-mercapto, 4-mercapto-benzoic acid, 4-mercapto benzoic acid, 4-thiobenzoic acid, p-mercaptobenzoic acid, benzoic acid, p-mercapto, para-mercaptobenzoate, paracarboxythiophenol
PubChem CID 95738
IUPAC Name 4-sulfanylbenzoic acid
SMILES OC(=O)C1=CC=C(S)C=C1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 223.0°C to 225.0°C
Assay Percent Range 88% min. (HPLC)
Packaging Glass bottle
Linear Formula HSC6H4CO2H
Quantity 25 g
Formula Weight 154.19
Percent Purity 90%
Chemical Name or Material 4-Mercaptobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.