missing translation for 'onlineSavingsMsg'
Learn More

4-Mercaptobenzoic Acid, Technical Grade, TRC

Product Code. 30179166
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179166 1 g 1g
30179652 25 g 25g
30179299 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179166 Supplier Toronto Research Chemicals Supplier No. TRCM2525001G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1074-36-8
Molecular Formula C7 H6 O2 S
Molecular Weight (g/mol) 154.19
IUPAC Name 4-sulfanylbenzoic acid
SMILES OC(=O)c1ccc(S)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H6O2S/c8-7(9)5-1-3-6(10)4-2-5/h1-4,10H,(H,8,9)
Formula Weight 154.0089
Chemical Name or Material 4-Mercaptobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.