missing translation for 'onlineSavingsMsg'
Learn More

4-Mercaptobenzoic Acid, TRC

Product Code. 30179502
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179502 1 g 1g
30179029 5 g 5g
30179126 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179502 Supplier Toronto Research Chemicals Supplier No. TRCM2524801G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1074-36-8
Molecular Formula C7 H6 O2 S
Molecular Weight (g/mol) 154.19
IUPAC Name 4-sulfanylbenzoic acid
SMILES OC(=O)c1ccc(S)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H6O2S/c8-7(9)5-1-3-6(10)4-2-5/h1-4,10H,(H,8,9)
Formula Weight 154.0089
Chemical Name or Material 4-Mercaptobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.