missing translation for 'onlineSavingsMsg'
Learn More

4-Mercaptophenol, 98%

Product Code. 10273052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10273052 25 g Glass bottle 25g
10603393 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10273052 Supplier Thermo Scientific Acros Supplier No. 370630250
ChemsChrom Mega Deals
Use promo code 27993
Ver todas las promociones
 Restricted Item
Estimated Shipment: 30-11-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 637-89-8 | C6H6OS | 126.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 637-89-8
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.17
MDL Number MFCD00004850
InChI Key BXAVKNRWVKUTLY-UHFFFAOYSA-N
Synonym 4-mercaptophenol, 4-hydroxythiophenol, 4-hydroxybenzenethiol, p-hydroxythiophenol, phenol, 4-mercapto, p-mercaptophenol, thiohydroquinone, monothiohydroquinone, 4-hydroxy thiophenol, hydroquinone, monothio
PubChem CID 240147
IUPAC Name 4-sulfanylphenol
SMILES OC1=CC=C(S)C=C1

Especificaciones

CAS Min % 98.5
CAS Max % 100.0
Melting Point 33°C to 35°C
Color Colorless to Yellow
Boiling Point 149°C to 150°C (25.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula SHC6H4OH
Quantity 25 g
Beilstein 06, IV, 5790
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,alkali solution,concentrated,sulfuric acid
Formula Weight 126.18
Percent Purity 99%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-Mercaptophenol, 99%
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.