missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxy-2'-nitroacetanilide, 98+%

Product Code. p-7044046
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11306856

Brand: Thermo Scientific Alfa Aesar L04218.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 119-81-3 | C9H10N2O4 | 210.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-81-3
Molecular Formula C9H10N2O4
Molecular Weight (g/mol) 210.189
MDL Number MFCD00017018
InChI Key QGEGALJODPBPGR-UHFFFAOYSA-N
Synonym n-4-methoxy-2-nitrophenyl acetamide, 4'-methoxy-2'-nitroacetanilide, 4-methoxy-2-nitroacetanilide, 2'-nitro-p-acetanisidide, acetamide, n-4-methoxy-2-nitrophenyl, 2-nitro-p-acetanisidide, p-acetanisidide, 2-nitro, 4-acetamido-3-nitroanisole, 2-nitro-4-methoxyacetanilide, p-acetanisidide, 2'-nitro
PubChem CID 67075
IUPAC Name N-(4-methoxy-2-nitrophenyl)acetamide
SMILES CC(=O)NC1=C(C=C(C=C1)OC)[N+](=O)[O-]

Specifications

Melting Point 117°C to 118°C
Quantity 5 g
Beilstein 2809678
Formula Weight 210.19
Percent Purity ≥98%
Chemical Name or Material 4'-Methoxy-2'-nitroacetanilide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.