missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-nitroaniline, 97%

Product Code. 10615752 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10615752 100 g Glass bottle 100g
10061270 25 g Glass bottle 25g
10153270 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10615752 Supplier Thermo Scientific Acros Supplier No. 125961000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 96-96-8 | C7H8N2O3 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 96-96-8
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.15
MDL Number MFCD00007152
InChI Key QFMJFXFXQAFGBO-UHFFFAOYSA-N
Synonym 2-nitro-4-methoxyaniline, 4-amino-3-nitroanisole, fast bordeaux gp, 2-nitro-p-anisidine, azobase nas, devol bordeaux b, bordeaux gp base, bordeaux gp salt, fast bordeaux gp base, fast bordeaux gp salt
PubChem CID 66793
ChEBI CHEBI:48973
IUPAC Name 4-methoxy-2-nitroaniline
SMILES COC1=CC(=C(C=C1)N)[N+](=O)[O-]

Specifica

CAS Min % 96.0
CAS Max % 100.0
Melting Point 123.0°C to 126.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3OC6H3(NO2)NH2
Quantity 100 g
Beilstein 13, 521
Solubility Information Solubility in water: slightly soluble
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material 4-Methoxy-2-nitroaniline, 98%
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons
Identificativo prodotto
  • 4-Methoxy-2-nitroaniline
Avvertenza
  • Danger
Categoria di pericolo
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Specific target organ toxicity after repeated exposure Category 2
Indicazioni di pericolo
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
Consigli di prudenza
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.