missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-nitroaniline, 99%

Product Code. p-7081625
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11419163

Brand: Thermo Scientific Alfa Aesar A11673.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-96-8 | C7H8N2O3 | 168.152 g/mol

4-Methoxy-2-nitroaniline is an aniline compound used in the synthesis of dyes and pigments. It has also been shown to act as a molluscicidal agent and has been used in hemolytic anemia studies. Also used as an intermediate for the synthesis of dyes, pigments and other chemical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-2-nitroaniline is an aniline compound used in the synthesis of dyes and pigments. It has also been shown to act as a molluscicidal agent and has been used in hemolytic anemia studies. Also used as an intermediate for the synthesis of dyes, pigments and other chemical compounds.

Solubility
Slightly soluble in water. Soluble in hot methanol almost transparency.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-96-8
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.152
MDL Number MFCD00007152
InChI Key QFMJFXFXQAFGBO-UHFFFAOYSA-N
Synonym 2-nitro-4-methoxyaniline, 4-amino-3-nitroanisole, fast bordeaux gp, 2-nitro-p-anisidine, azobase nas, devol bordeaux b, bordeaux gp base, bordeaux gp salt, fast bordeaux gp base, fast bordeaux gp salt
PubChem CID 66793
ChEBI CHEBI:48973
IUPAC Name 4-methoxy-2-nitroaniline
SMILES COC1=CC(=C(C=C1)N)[N+](=O)[O-]

Specifications

Melting Point 126°C to 128°C
Quantity 100 g
UN Number UN2811
Beilstein 880318
Solubility Information Slightly soluble in water. Soluble in hot methanol almost transparency.
Formula Weight 168.15
Percent Purity 99%
Chemical Name or Material 4-Methoxy-2-nitroaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.