missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-(trifluoromethyl)benzeneboronic acid

Product Code. 11476110
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476110 1 g 1g
11486110 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476110 Supplier Thermo Scientific Alfa Aesar Supplier No. H27062.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 313546-16-6 | C8H8BF3O3 | 219.954 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313546-16-6
Molecular Formula C8H8BF3O3
Molecular Weight (g/mol) 219.954
MDL Number MFCD03095346
InChI Key ZBCRZEJNAADYKG-UHFFFAOYSA-N
Synonym 4-methoxy-2-trifluoromethyl phenylboronic acid, 4-methoxy-2-trifluoromethyl benzeneboronic acid, 4-methoxy-2-trifluoromethyl phenyl boronic acid, 4-methoxy-2-trifluoromethyl-phenylboronic acid, 4-methoxy-2, pubchem1868, acmc-1aefl, ablock ab-13-0076
PubChem CID 12109460
IUPAC Name [4-methoxy-2-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=C(C=C(C=C1)OC)C(F)(F)F)(O)O

Specifications

Melting Point 168°C to 172°C
Quantity 1 g
Formula Weight 219.95
Chemical Name or Material 4-Methoxy-2-(trifluoromethyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.