missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-buten-2-one, 90%, tech.

Product Code. 10103540 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
This item is not returnable. View return policy

Product Code. 10103540

Brand: Thermo Scientific Acros 153390100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4652-27-1 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4652-27-1
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008778,MFCD00008778,MFCD00008778
InChI Key VLLHEPHWWIDUSS-ARJAWSKDSA-N
Synonym z-4-methoxybut-3-en-2-one, 3z-4-methoxybut-3-en-2-one, 3-buten-2-one, 4-methoxy-, 3z, z-4-methoxy-3-butene-2-one
PubChem CID 643837
IUPAC Name (Z)-4-methoxybut-3-en-2-one
SMILES CO\C=C/C(C)=O

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Yellow
Density 0.9800g/mL
Boiling Point 200°C
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula CH3OCH=CHCOCH3
Refractive Index 1.4650 to 1.469
Quantity 10 mL
Beilstein 01, IV, 4080
Fieser 10,258
Specific Gravity 0.98
Solubility Information Solubility in water: insoluble
Formula Weight 100.12
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material 4-Methoxy-3-buten-2-one, tech., 90%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxy-3-buten-2-one
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.