missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-methylbenzyl alcohol

Product Code. 11499520
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11499520

Brand: Thermo Scientific Alfa Aesar H50344.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 114787-91-6 | C9H12O2 | 152.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114787-91-6
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00060357
InChI Key UVIIQYUUEVQCOR-UHFFFAOYSA-N
Synonym 4-methoxy-3-methylbenzyl alcohol, 4-methoxy-3-methylphenyl methanol, 4-methoxy-3-methylbenzyl pound inverted question markalcohol, pubchem24130, acmc-1c48b, 3-methyl-4-methoxybenzyl alcohol, 4-methoxy-3-methyl-benzyl alcohol, benzenemethanol,4-methoxy-3-methyl, 4-methoxy-3-methylphenyl methan-1-ol
PubChem CID 14457544
IUPAC Name (4-methoxy-3-methylphenyl)methanol
SMILES CC1=C(C=CC(=C1)CO)OC

Specifications

Quantity 250 mg
Formula Weight 152.19
Chemical Name or Material 4-Methoxy-3-methylbenzyl alcohol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.