missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-nitrobenzoic acid, 98%, Thermo Scientific™

Product Code. 10174010 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
Product Code. Quantity Packaging unitSize
10174010 25 g Glass bottle 25g
10702811 100 g Glass bottle 100g
2 options
This item is not returnable. View return policy

Product Code. 10174010

Brand: Thermo Scientific Acros 187830250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 89-41-8
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.15
MDL Number MFCD00007256
InChI Key ANXBDAFDZSXOPQ-UHFFFAOYSA-N
Synonym 4-methoxy-3-nitrobenzoic acid, 3-nitro-4-methoxybenzoic acid, 3-nitro-p-anisic acid, benzoic acid, 4-methoxy-3-nitro, unii-dd71o31olz, p-anisic acid, 3-nitro, dd71o31olz, 4-methoxy-3-nitro benzoic acid, 4-methoxy-3-nitro-benzoic acid, 4-methoxy-3-nitrobenzoicacid
PubChem CID 66640
SMILES COC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 189°C to 194°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3OC6H3(NO2)CO2H
Quantity 25 g
Beilstein 10, 181
Formula Weight 197.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-3-nitrobenzoic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.