missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific™

Product Code. 15437555
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437555 250 mg 250mg
11382119 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437555 Supplier Thermo Scientific Alfa Aesar Supplier No. H26128.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Slightly soluble in water.

Notes
Air Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50823-87-5
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD01091012
InChI Key UMZQFCFPWAFMRL-UHFFFAOYSA-N
Synonym 4-methoxy-3-trifluoromethyl benzaldehyde, 3-trifluoromethyl-4-methoxybenzaldehyde, pubchem4247, acmc-1aqw0, 3-trifluoromethyl-p-anisaldehyde, benzaldehyde, 4-methoxy-3-trifluoromethyl
PubChem CID 2775291
IUPAC Name 4-methoxy-3-(trifluoromethyl)benzaldehyde
SMILES COC1=C(C=C(C=C1)C=O)C(F)(F)F

Specifications

Melting Point 38°C to 40°C
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 204.15
Percent Purity 98%
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.