missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™

Product Code. 11382459
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382459 1 g 1g
11392459 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382459 Supplier Thermo Scientific Alfa Aesar Supplier No. H26606.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
As organic intermediate, pharmaceutical intermediate. In chemical synthesis.

Solubility
Sparingly Soluble (0.16 g/L) (25°C).

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 261951-89-7
Molecular Formula C9H8BrF3O
Molecular Weight (g/mol) 269.061
MDL Number MFCD01631578
InChI Key FZICFAROPYFJMF-UHFFFAOYSA-N
Synonym 4-methoxy-3-trifluoromethyl benzyl bromide, 4-bromomethyl-1-methoxy-2-trifluoromethyl benzene, benzene, 4-bromomethyl-1-methoxy-2-trifluoromethyl, 4-bromomethyl-1-methoxy-2-trifluoromethyl-benzene, 4-bromomethyl-2-trifluoromethyl-1-methoxybenzene, acmc-20amfh, pubchem4655, 4-methoxy-3-trifluoromethyl benzylbromide
PubChem CID 2775302
IUPAC Name 4-(bromomethyl)-1-methoxy-2-(trifluoromethyl)benzene
SMILES COC1=C(C=C(C=C1)CBr)C(F)(F)F

Especificaciones

Melting Point 75°C to 79°C
Quantity 1 g
UN Number UN3261
Solubility Information Sparingly Soluble (0.16g/L) (25°C).
Formula Weight 269.06
Percent Purity 97%
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzyl bromide
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.