missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-4'-nitrobenzophenone, 97%

Product Code. 11495367
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495367 1 g 1g
11405377 5 g 5g
11415377 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495367 Supplier Thermo Scientific Alfa Aesar Supplier No. B22720.03
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1151-94-6 | C14H11NO4 | 257.25 g/mol

4-Methoxy-4'-nitrobenzophenone is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-4′-nitrobenzophenone is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1151-94-6
Molecular Formula C14H11NO4
Molecular Weight (g/mol) 257.25
MDL Number MFCD00178348
InChI Key DXVSAELNVPXMSQ-UHFFFAOYSA-N
Synonym 4-methoxy-4'-nitrobenzophenone, 4-methoxyphenyl 4-nitrophenyl methanone, 4-methoxyphenyl-4-nitrophenyl methanone, methanone, 4-methoxyphenyl 4-nitrophenyl, 4-methoxy-phenyl-4-nitro-phenyl-methanone, acmc-1buw2, 4-methoxy-4-nitrobenzophenone, 4-methoxy-4?-nitrobenzophenone, 4-methoxy-4/'-nitrobenzophenone
PubChem CID 347772
IUPAC Name (4-methoxyphenyl)-(4-nitrophenyl)methanone
SMILES COC1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 125°C to 127°C
Quantity 1 g
Formula Weight 257.25
Percent Purity 97%
Chemical Name or Material 4-Methoxy-4'-nitrobenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.