missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide, 97%, Thermo Scientific™

Product Code. 15493716
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493716 250 mg 250mg
15483716 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493716 Supplier Thermo Scientific Alfa Aesar Supplier No. H57808.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide is involved in long-chain fatty acid stimulated GLP-1 secretion and has been implicated in glucose homeostasis, adiposity, and regulation of the anti-inflammatory response. It enhances glucose-stimulated insulin secretion in MIN6 cells. It induces intracellular calcium accumulation in U2OS cells; this activity is inhibited by AH 7614.

Solubility
Soluble to 100 mM in DMSO and to 50 mM in ethanol.

Notes
Store away from oxidizing agents. Store in a dry, cool conditions in a well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 349085-82-1
Molecular Formula C16H19NO3S
Molecular Weight (g/mol) 305.392
MDL Number MFCD02007969
InChI Key FQUAFMNPXPXOJE-UHFFFAOYSA-N
Synonym 4-methoxy-n-2,4,6-trimethylphenyl benzenesulfonamide, cbmicro_012617, n-mesityl-4-methoxy-1-benzenesulfonamide, gsk137647a hplc, 4-methoxyphenyl sulfonyl 2,4,6-trimethylphenyl amine, 4-methoxy-n-2,4,6-trimethylphenyl benzene-1-sulfonamide
PubChem CID 743974
IUPAC Name 4-methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide
SMILES CC1=CC(=C(C(=C1)C)NS(=O)(=O)C2=CC=C(C=C2)OC)C

Specifications

Quantity 250 mg
Beilstein 23102756
Solubility Information Soluble to 100 mM in DMSO and to 50 mM in ethanol.
Formula Weight 305.39
Percent Purity 97%
Chemical Name or Material 4-Methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.