missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-N-methylbenzylamine, 95%

Product Code. 15254894 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15254894 1 g Glass bottle 1g
15274894 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15254894 Supplier Thermo Scientific Acros Supplier No. 451730010
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 702-24-9 | C9H13NO | 151.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 702-24-9
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
InChI Key AIJFPNKGGAPZFJ-UHFFFAOYSA-N
Synonym 4-methoxy-n-methylbenzylamine, n-4-methoxybenzyl-n-methylamine, n-methyl-4-methoxybenzylamine, 1-4-methoxyphenyl-n-methylmethanamine, 4-methoxyphenyl methyl methyl amine, benzenemethanamine, 4-methoxy-n-methyl, 4-methoxy-benzyl-methyl-amine, 4-methoxyphenyl-n-methylmethanamine, n-methyl-p-methoxybenzylamine, 1-4-methoxyphenyl-n-methyl-methanamine
PubChem CID 485407
IUPAC Name 1-(4-methoxyphenyl)-N-methylmethanamine
SMILES CNCC1=CC=C(C=C1)OC

Specifications

Density 1.0080g/mL
Boiling Point 222.0°C
Flash Point 109°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CH2NCH3
Quantity 1 g
Specific Gravity 1.008
Solubility Information Solubility in water: slightly soluble
Formula Weight 151.21
Percent Purity 95%
Chemical Name or Material 4-Methoxy-N-methylbenzylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-(4-Methoxybenzyl)-N-methylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.