missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxyacetanilide, 99+%, ACROS Organics™

Product Code. 10790551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
50g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10790551 50g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10790551 Supplier Thermo Scientific Acros Supplier No. 249710500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 51-66-1
CAS Min % 99.0
CAS Max % 100.0
Melting Point 129°C to 132°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C9H11NO2
Linear Formula CH3C6H4NHC(O)CH3
MDL Number MFCD00014963
Quantity 50 g
Merck Index 15, 50
Synonym n-4-methoxyphenyl acetamide, 4'-methoxyacetanilide, p-methoxyacetanilide, p-acetanisidide, p-acetanisidine, methacetin, 4-methoxyacetanilide, aceto-p-anisidide, n-acetyl-p-anisidine, acetamide, n-4-methoxyphenyl
InChI Key XVAIDCNLVLTVFM-UHFFFAOYSA-N
SMILES CC(=O)NC1=CC=C(C=C1)OC
IUPAC Name N-(4-methoxyphenyl)acetamide
Molecular Weight (g/mol) 165.192
PubChem CID 5827
Formula Weight 165.19
Percent Purity ≥99.0%
Physical Form Crystalline Powder
Chemical Name or Material 4'-Methoxyacetanilide, 99+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.