missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzhydrazide, 98+%

Product Code. 11479923
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479923 50 g 50g
11469923 10 g 10g
11489923 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479923 Supplier Thermo Scientific Alfa Aesar Supplier No. A13015.18
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3290-99-1 | C8H10N2O2 | 166.18 g/mol

4-Methoxybenzhydrazide is used as pharmaceutical intermediates and it is also involved in a variety of organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzhydrazide is used as pharmaceutical intermediates and it is also involved in a variety of organic synthesis.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3290-99-1
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00017073
InChI Key REKQLYUAUXYJSZ-UHFFFAOYSA-N
Synonym 4-methoxybenzhydrazide, p-anisohydrazide, p-anisic hydrazide, p-anisoylhydrazine, 4-methoxybenzoic acid hydrazide, 4-methoxybenzoylhydrazine, anisoylhydrazine, anisic hydrazide, 4-methoxybenzene-1-carbohydrazide, p-anisic acid, hydrazide
PubChem CID 76792
IUPAC Name 4-methoxybenzohydrazide
SMILES COC1=CC=C(C=C1)C(=O)NN

Specifications

Melting Point 136°C to 140°C
Quantity 50 g
Beilstein 389017
Solubility Information Slightly soluble in water.
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 4-Methoxybenzhydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.