missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzhydrol, 98+%

Product Code. 11310064
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11310064 25 g 25g
11300064 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11310064 Supplier Thermo Scientific Alfa Aesar Supplier No. L00468.14
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 720-44-5 | C14H14O2 | 214.264 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 720-44-5
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.264
MDL Number MFCD00014398
InChI Key BEGZWXVLBIZFKQ-UHFFFAOYSA-N
Synonym 4-methoxybenzhydrol, 4-methoxyphenyl phenyl methanol, 4-methoxybenzhydryl alcohol, benzenemethanol, 4-methoxy-.alpha.-phenyl, 4-methoxyphenyl phenylmethan-1-ol, 4-methoxy-phenyl-phenyl-methanol, mamp-oh resin, 4-methoxybenzohydrol, acmc-209olf, p-methoxybenzhydryl alcohol
PubChem CID 95375
IUPAC Name (4-methoxyphenyl)-phenylmethanol
SMILES COC1=CC=C(C=C1)C(C2=CC=CC=C2)O

Specifications

Melting Point 65°C to 68°C
Quantity 25 g
Beilstein 3202445
Formula Weight 214.26
Percent Purity ≥98%
Chemical Name or Material 4-Methoxybenzhydrol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.