missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzylamine, 98%

Product Code. 11975271 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass Bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11975271 500 mL Glass Bottle 500mL
10051120 25 mL Glass Bottle 25mL
10193410 100 mL Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11975271 Supplier Thermo Scientific Acros Supplier No. 125785000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2393-23-9 | C8H11NO | 137.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2393-23-9
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
InChI Key IDPURXSQCKYKIJ-UHFFFAOYSA-N
Synonym 4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine
PubChem CID 75452
ChEBI CHEBI:49837
IUPAC Name (4-methoxyphenyl)methanamine
SMILES COC1=CC=C(C=C1)CN

Specifications

Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 236.0°C to 237.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3OC6H4CH2NH2
Refractive Index 1.5460 to 1.5480
Quantity 500 mL
Beilstein 13,606
Specific Gravity 1.05
Formula Weight 137.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Methoxybenzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxybenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.