missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzylchloride, 98%, stabilized

Product Code. 10449052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10449052 25 g 25g
10535481 5 g 5g
15264694 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10449052 Supplier Thermo Scientific Acros Supplier No. 294170250
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 824-94-2 | C8H9ClO | 156.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824-94-2 , 584-08-7
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00000915
InChI Key MOHYOXXOKFQHDC-UHFFFAOYSA-N
Synonym 4-methoxybenzyl chloride, 4-methoxybenzylchloride, 1-chloromethyl-4-methoxybenzene, p-methoxybenzyl chloride, p-anisyl chloride, alpha-chloro-4-methoxytoluene, p-chloromethyl anisole, benzene, 1-chloromethyl-4-methoxy, 4-chloromethyl anisole, anisole, p-chloromethyl
PubChem CID 69993
IUPAC Name 1-(chloromethyl)-4-methoxybenzene
SMILES COC1=CC=C(C=C1)CCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -1.0°C
Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 117.0°C to 118.0°C (14.0 mmHg)
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5470 to 1.5490
Quantity 25 g
Specific Gravity 1.15
Solubility Information Solubility in water: slowly decomposes
Stabilizer 1% K2CO3
Formula Weight 156.61
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Methoxybenzylchloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxybenzylchloride, stabilized
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.