missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybiphenyl, 98+%

Product Code. 11354524
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11354524 5 g 5g
11364524 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354524 Supplier Thermo Scientific Alfa Aesar Supplier No. L01260.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 613-37-6 | C13H12O | 184.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-37-6
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.24
MDL Number MFCD00014897
InChI Key RHDYQUZYHZWTCI-UHFFFAOYSA-N
Synonym 4-methoxybiphenyl, 4-phenylanisole, p-methoxybiphenyl, 1,1'-biphenyl, 4-methoxy, 4-methoxy-1,1'-biphenyl, anisole, p-phenyl, p-phenylanisole, 4-methoxy-biphenyl, biphenyl, 4-methoxy, 1-methoxy-4-phenyl-benzene
PubChem CID 11943
IUPAC Name 1-methoxy-4-phenylbenzene
SMILES COC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 86°C to 90°C
Boiling Point 157°C (10 mmHg)
Quantity 5 g
Beilstein 2043157
Formula Weight 184.24
Percent Purity ≥98%
Chemical Name or Material 4-Methoxybiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.