missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxycarbonyl-2-nitrobenzeneboronic acid, 97%

Product Code. 15449637
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449637 5 g 5g
15439637 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15449637 Supplier Thermo Scientific Alfa Aesar Supplier No. H52863.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 85107-55-7 | C8H8BNO6 | 224.963 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85107-55-7
Molecular Formula C8H8BNO6
Molecular Weight (g/mol) 224.963
MDL Number MFCD01632202
InChI Key SFEJGHJCGQNFQC-UHFFFAOYSA-N
Synonym 4-methoxycarbonyl-2-nitrophenyl boronic acid, 4-methoxycarbonyl-2-nitrophenylboronic acid, 4-methoxycarbonyl-2-nitrobenzeneboronic acid, methyl 4-dihydroxyboranyl-3-nitrobenzoate, 2-nitro-4-methoxycarbonyl phenylboronic acid, 4-methoxycarbonyl-2-nitro-phenyl boronic acid, methyl 4-borono-3-nitrobenzoate, acmc-209q4d
PubChem CID 2773493
IUPAC Name (4-methoxycarbonyl-2-nitrophenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)C(=O)OC)[N+](=O)[O-])(O)O

Specifications

Melting Point 160°C to 171°C
Quantity 5 g
Formula Weight 224.96
Percent Purity 97%
Chemical Name or Material 4-Methoxycarbonyl-2-nitrobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.