missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxychalcone, 97%

Product Code. 11392096
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392096 25 g 25g
11382096 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392096 Supplier Thermo Scientific Alfa Aesar Supplier No. L03319.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 959-33-1 | C16H14O2 | 238.286 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 959-33-1
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
MDL Number MFCD00017179
InChI Key XUFXKBJMCRJATM-FMIVXFBMSA-N
Synonym 4-methoxychalcone, e-3-4-methoxyphenyl-1-phenylprop-2-en-1-one, 2e-3-4-methoxyphenyl-1-phenylprop-2-en-1-one, chalcone, 4-methoxy, 3-4-methoxyphenyl-1-phenyl-2-propen-1-one, 2-propen-1-one, 3-4-methoxyphenyl-1-phenyl, 2-propen-1-one, 3-4-methoxyphenyl-1-phenyl-, 2e, phenyl p-methoxystyryl ketone, 4'-methoxybenzylideneacetophenone, 3-4-methoxyphenyl-1-phenylprop-2-en-1-one
PubChem CID 641819
IUPAC Name (E)-3-(4-methoxyphenyl)-1-phenylprop-2-en-1-one
SMILES COC1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2

Specifications

Melting Point 73°C to 76°C
Quantity 25 g
Beilstein 978742
Formula Weight 238.29
Percent Purity 97%
Chemical Name or Material 4-Methoxychalcone
compliance-icons
Product Identifier
  • 4-Methoxychalcone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 4-Methoxychalcone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.