missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxychalcone, 97%

Product Code. 11304709
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304709 5 g 5g
11439687 25 g 25g
11449687 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11304709 Supplier Thermo Scientific Alfa Aesar Supplier No. B22158.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 959-23-9 | C16H14O2 | 238.286 g/mol

4-Methoxychalcone is used as a precursor to prepare 3-(4-methoxy-phenyl)-3-morpholin-4-yl-1-phenyl-propan-1-one by reacting with morpholine in presence of heptanes as solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxychalcone is used as a precursor to prepare 3-(4-methoxy-phenyl)-3-morpholin-4-yl-1-phenyl-propan-1-one by reacting with morpholine in presence of heptanes as solvent.

Solubility
Soluble in dichloromethane and methanol. Insoluble in water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible withstrong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 959-23-9
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
MDL Number MFCD00008407
InChI Key KJHHAPASNNVTSN-KPKJPENVSA-N
Synonym 4'-methoxychalcone, 2e-1-4-methoxyphenyl-3-phenylprop-2-en-1-one, 1-4-methoxyphenyl-3-phenylprop-2-en-1-one, chembl34398, 1-4-methoxyphenyl-3-phenyl-2-propen-1-one, e-1-4-methoxyphenyl-3-phenylprop-2-en-1-one, unii-bm45n45fiz, e-1-4-methoxy-phenyl-3-phenyl-propenone, 2-propen-1-one, 1-4-methoxyphenyl-3-phenyl-, 2e, 2-propen-1-one, 1-4-methoxyphenyl-3-phenyl
PubChem CID 641818
IUPAC Name (E)-1-(4-methoxyphenyl)-3-phenylprop-2-en-1-one
SMILES COC1=CC=C(C=C1)C(=O)C=CC2=CC=CC=C2

Specifications

Melting Point 101°C to 103°C
Quantity 5 g
Beilstein 644640
Solubility Information Soluble in dichloromethane and methanol. Insoluble in water.
Formula Weight 238.29
Percent Purity 97%
Chemical Name or Material 4'-Methoxychalcone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.