missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenoxyacetic acid, 98%

Product Code. 11487047
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487047 10 g 10g
11497047 50 g 50g
11407057 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487047 Supplier Thermo Scientific Alfa Aesar Supplier No. A14419.09
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1877-75-4 | C9H10O4 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1877-75-4
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00014360
InChI Key BHFSBJHPPFJCOS-UHFFFAOYSA-N
Synonym 4-methoxyphenoxyacetic acid, 2-4-methoxyphenoxy acetic acid, 4-methoxyphenoxy acetic acid, acetic acid, 4-methoxyphenoxy, p-methoxyphenoxyacetic acid, acetic acid, p-methoxyphenoxy, acetic acid, 2-4-methoxyphenoxy, p-methoxyphenoxy acetic acid, akos bb/0049, rarechem al bo 0321
PubChem CID 74649
IUPAC Name 2-(4-methoxyphenoxy)acetic acid
SMILES COC1=CC=C(OCC(O)=O)C=C1

Specifications

Melting Point 109°C to 113°C
Quantity 10 g
Beilstein 1874579
Formula Weight 182.18
Percent Purity 98%
Chemical Name or Material 4-Methoxyphenoxyacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.