missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl chloroformate, 98%

Product Code. 10741731 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10741731 5 g Glass Bottle 5g
10321244 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10741731 Supplier Thermo Scientific Acros Supplier No. 314570050
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7693-41-6 | C8H7ClO3 | 186.591 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7693-41-6
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00013258
InChI Key CCFSGQKTSBIIHG-UHFFFAOYSA-N
Synonym 4-methoxyphenyl chloroformate, carbonochloridic acid, 4-methoxyphenyl ester, 4-methoxyphenylchloroformate, chloroformic acid 4-methoxyphenyl ester, acmc-1bfyr, p-methoxyphenyl chloroformate, 4-methoxy-phenyl chloroformate, 4-methoxy phenyl chloroformate, 4-methoxyphenyl carbonochloridate
PubChem CID 82128
IUPAC Name (4-methoxyphenyl) carbonochloridate
SMILES COC1=CC=C(C=C1)OC(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.25g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula ClCOC6H4OCH3
Refractive Index 1.5220 to 1.5260 (20°C, 589nm)
Quantity 5 g
UN Number 3277
Formula Weight 186.59
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenyl chloroformate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.