missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl isocyanate, 99%

Product Code. 10657352 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10657352 5 g Glass bottle 5g
10733411 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10657352 Supplier Thermo Scientific Acros Supplier No. 218760050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5416-93-3 | C8H7NO2 | 149.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5416-93-3
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.15
MDL Number MFCD00002026
InChI Key FMDGXCSMDZMDHZ-UHFFFAOYSA-N
Synonym 4-methoxyphenyl isocyanate, 4-methoxyphenylisocyanate, benzene, 1-isocyanato-4-methoxy, p-anisyl isocyanate, p-methoxyphenyl isocyanate, isocyanic acid, p-methoxyphenyl ester, unii-c50o5096nc, 1-isocyanato-4-methoxy-benzene, 4-methoxybenzenisocyanate, 4-methoxyisocyanatobenzene
PubChem CID 79443
IUPAC Name 1-isocyanato-4-methoxybenzene
SMILES COC1=CC=C(C=C1)N=C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.1500g/mL
Boiling Point 106°C to 110°C (16.0 mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4NCO
Refractive Index 1.5450 to 1.547
Quantity 5 g
Beilstein 13, 487
Specific Gravity 1.15
Formula Weight 149.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenyl isocyanate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxyphenyl isocyanate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.