missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 10529952 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10529952 10 g 10g
10577294 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10529952 Supplier Thermo Scientific Supplier No. TL00326EA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2284-20-0
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00011676
InChI Key VRPQCVLBOZOYCG-UHFFFAOYSA-N
Synonym 4-methoxyphenyl isothiocyanate, 4-methoxyphenylisothiocyanate, 1-isothiocyanato-4-methoxy-benzene, p-methoxyphenyl isothiocyanate, benzene, 1-isothiocyanato-4-methoxy, isothiocyanic acid 4-methoxyphenyl ester, 4-methoxybenzenisothiocyanate, 4-isothiocyanatoanisole, acmc-1ccqx, p-methoxyphenylisothiocyanate
PubChem CID 75293
IUPAC Name 1-isothiocyanato-4-methoxybenzene
SMILES COC1=CC=C(C=C1)N=C=S

Specifications

CAS Min % 97.0
Density 1.1900g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3OC6H4NCS
Quantity 10 g
Formula Weight 165.22
Percent Purity 97%
Chemical Name or Material 4-Methoxyphenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.